missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLPX Recombinant Rabbit Monoclonal Antibody (23GB1745), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | CLPX |
|---|---|
| Clone | 23GB1745 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089643
|
Invitrogen™
MA552590 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087692
|
Invitrogen™
MA552591 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLPX Recombinant Monoclonal antibody specifically detects CLPX in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| CLPX | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O76031, Q5U2U0, Q9JHS4 | |
| 10845, 270166, 300786 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1745 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ATP-dependent Clp protease ATP-binding subunit clpX-like, mitochondrial; AU014732; caseinolytic mitochondrial matrix peptidase chaperone subunit; caseinolytic peptidase X; caseinolytic protease X; CLPX; ClpX caseinolytic peptidase X homolog; ClpX caseinolytic protease X homolog; E330029I21; energy-dependent regulator of proteolysis | |
| A synthesized peptide derived from human CLPX (600-633AA). | |
| Primary | |
| -20°C | |
| CLPX |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title