missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLS1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 193.00 - € 483.00
Specifications
| Antigen | CLS1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18373417
|
Novus Biologicals
NBP3-03235-20UL |
20 μL |
€ 193.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18033164
|
Novus Biologicals
NBP3-03235-100UL |
100 μL |
€ 483.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLS1 Polyclonal specifically detects CLS1 in Mouse samples. It is validated for Western Blot.Specifications
| CLS1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 54675 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| cardiolipin synthase, cardiolipin synthase 1, CLS, dJ967N21.6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human CLS1 (NP_061968.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title