Learn More
CLSTN2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21436-1-AP-150UL
 View more versions of this product
Product Code. 18039465
  • € 410.00 / 150µL
Description
The CLSTN2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CLSTN2. This antibody recognizes human, mouse antigen. The CLSTN2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CLSTN2
300 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
ALC GAMMA, Alcadein gamma, alcagamma, calsyntenin 2, CLSTN2, CS2, CSTN2
150 μL
64084
Store at -20°C. Stable for one year after shipment.