Learn More
CLSTN2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21436-1-AP-20UL
Description
The CLSTN2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CLSTN2. This antibody recognizes human, mouse antigen. The CLSTN2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CLSTN2 | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| ALC GAMMA, Alcadein gamma, alcagamma, calsyntenin 2, CLSTN2, CS2, CSTN2 | |
| 20 μL | |
| 64084 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |