Learn More
Invitrogen™ CLTCL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568407
This item is not returnable.
View return policy
Description
CLTCL1 Polyclonal Antibody for IHC (P)
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
Specifications
| CLTCL1 | |
| Polyclonal | |
| Unconjugated | |
| CLTCL1 | |
| CHC22; clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 22; clathrin, heavy chain-like 1; Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; CLH22; CLH-22; CLTCL; CLTCL1; CLTD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8218 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P53675 | |
| CLTCL1 | |
| Synthetic peptide corresponding to residues near the C terminal of human CLTCL1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |