missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Clusterin Alpha Recombinant Rabbit Monoclonal Antibody (5I2Q1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551294
This item is not returnable.
View return policy
Description
Immunogen sequence: STNNPSQAKL RRELDESLQV AERLTRKYNE LLKSYQWKML NTSSLLEQLN EQFNWVSRLA NLTQGEDQYY LRVTTVASHT SDSDVPSGVT EVVVKLFDSD PITVTVPVEV SRKNPKFMET VAEKALQEYR KKHREE.
Isoform 1 of Clusterin prevents stress-induced aggregation of blood plasma proteins. Inhibits formation of amyloid fibrils by APP, APOC2, B2M, CALCA, CSN3, SNCA and aggregation-prone LYZ variants (in vitro). Does not require ATP. Maintains partially unfolded proteins in a state appropriate for subsequent refolding by other chaperones, such as HSPA8/HSC70. Does not refold proteins by itself. Binding to cell surface receptors triggers internalization of the chaperone-client complex and subsequent lysosomal or proteasomal degradation. Intracellular isoforms interact with ubiquitin and SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complexes and promote the ubiquitination and subsequent proteasomal degradation of target proteins. Promotes proteasomal degradation of COMMD1 and IKBKB. Modulates NF-kappa-B transcriptional activity. Nuclear isoforms promote apoptosis. Mitochondrial isoforms suppress BAX-dependent release of cytochrome c into the cytoplasm and inhibit apoptosis. Plays a role in the regulation of cell proliferation.
Specifications
| Clusterin Alpha | |
| Recombinant Monoclonal | |
| 0.98 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P10909, Q06890 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1191, 12759 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5I2Q1 | |
| Unconjugated | |
| CLU | |
| CLU | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 314-449 of human Clusterin alpha chain (NP_001822.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction