missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMAS, Mouse anti-Human, Clone: 5A2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00055907-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: EGYDSVFSVVRRHQFRWSEIQKGVREVTEPLNLNPAKRPRRQDWDGELYENGSFYFAKRHLIEMGYLQGGKMAYYEMRAEHSVDIDVDIDWPIAEQRVLRSpecifications
| CMAS | |
| Monoclonal | |
| Unconjugated | |
| cytidine monophosphate N-acetylneuraminic acid synthetase | |
| - | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 5A2 | |
| In 1x PBS, pH 7.4 | |
| NM_018686 | |
| CMAS | |
| CMAS (NP_061156, 164 a.a. ∼ 263 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 55907 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction