missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMC1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97239DL594
This item is not returnable.
View return policy
Description
CMC1 Polyclonal antibody specifically detects CMC1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CMC1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| C3orf68, chromosome 3 open reading frame 68, cmc1p, COX assembly mitochondrial protein homolog, COX assembly mitochondrial protein homolog (S. cerevisiae), CX9C mitochondrial protein required for full expression of COX 1, DKFZp779D0833, MGC61571, mitochondrial metallochaperone-like protein | |
| Produced in rabbits immunized with E. coli-derived Human CMC1 fragment. | |
| 0.1 mL | |
| metabolism | |
| 152100 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction