missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CMIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566862
This item is not returnable.
View return policy
Description
Immunogen sequence: RDQWFHSLQWK KKIYKYKKVL SNPSRWEVVL KEIRTLVDMA LTSPLQDDSI NQAPLEIVSK LLSENTNLTT QEHENIIVAI APLLENNH Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
The munc-18 interacting protein (Mint) protein family is a group of evolutionarily conserved adaptor proteins that function in membrane transport and organization. In mammals, there exist three Mint isoforms, Mint1, 2, and 3. Although there is little amino acid sequence conservation in the amino-terminal half, the carboxy-terminal half of these proteins is highly conserved. Within this conserved portion there exists a phosphotyrosine-binding (PTB) and a PSD-95/DLG-A/ZO-1 (PDZ) domain, which function as protein interaction modules. Mint1 and 2 appear to be expressed exclusively in the brain and are found to bind to Munc18, an essential component of the synaptic vesicle fusion machinery. Mint3 is ubiquitously expressed in all tissues and is expressed at the lowest levels in the brain and testis. Studies show that mint3 does not interact with munc-18. Mint3 has been found to interact with the Alzheimer's Disease-related amyloid precursor protein (APP) and does so through its PTB and PDZ domains. It has been suggested that mint3 links APP to other transport machinery components, thereby regulating it transport, endocytosis, and metabolism. Abnormal APP metabolism has been shown to be the cause of an early-onset type of Alzheimer's disease.
Specifications
| CMIP | |
| Polyclonal | |
| Unconjugated | |
| CMIP | |
| 4933407C03Rik; 5830471E12Rik; AI314065; AI464126; c-Maf inducing protein; c-Maf-inducing protein; Cmip; c-Mip; Kiaa1694; RGD1306101; TCMIP; Tc-Mip; Truncated c-Maf-inducing protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IY22 | |
| CMIP | |
| Recombinant Human CMIP. Recombinant protein control fragment (Product #RP-105403). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction