missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMP kinase Antibody (07), Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 326.00
Specifications
| Antigen | CMP kinase |
|---|---|
| Clone | 07 |
| Dilution | ELISA 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:20-1:100 |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679877
|
Novus Biologicals
NBP3-06538-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625096
|
Novus Biologicals
NBP3-06538-50ul |
50 μg |
€ 220.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CMP kinase Monoclonal antibody specifically detects CMP kinase in Human samples. It is validated for ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| CMP kinase | |
| ELISA 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:20-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CMKEC 2.7.4.14, CMPKUMKUMP-CMP kinase, cytidine monophosphate (UMP-CMP) kinase 1, cytosolic, Cytidine monophosphate kinase, Cytidylate kinaseUMP/CMP kinase, Deoxycytidylate kinase, EC 2.7.4, UMP/CMPK, UMP-CMPK, UMPKUCK, Uridine monophosphate kinase, Uridine monophosphate/cytidine monophosphate kinase | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CMP kinase (Uniprot#: P30085; Met1-Gly196). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 07 | |
| ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Proteases & Other Enzymes | |
| 0.2 um filtered solution in PBS | |
| 51727 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title