missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMP kinase Antibody (08), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06539-50ul
This item is not returnable.
View return policy
Description
CMP kinase Monoclonal antibody specifically detects CMP kinase in Human samples. It is validated for Western Blot
Specifications
| CMP kinase | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 08 | |
| Western Blot 1:500-1:2000 | |
| CMKEC 2.7.4.14, CMPKUMKUMP-CMP kinase, cytidine monophosphate (UMP-CMP) kinase 1, cytosolic, Cytidine monophosphate kinase, Cytidylate kinaseUMP/CMP kinase, Deoxycytidylate kinase, EC 2.7.4, UMP/CMPK, UMP-CMPK, UMPKUCK, Uridine monophosphate kinase, Uridine monophosphate/cytidine monophosphate kinase | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CMP kinase (Uniprot#: P30085; Met1-Gly196). | |
| 50 μg | |
| Proteases & Other Enzymes | |
| 51727 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction