missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CMTM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582489
This item is not returnable.
View return policy
Description
Immunogen sequence: NDVAKFLKQG DSADETTAHK TEEENSDSDS Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 83%.
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies of signaling molecules. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. Alternatively spliced transcript variants containing different 5' UTRs, but encoding the same protein, have been identified.
Specifications
| CMTM3 | |
| Polyclonal | |
| Unconjugated | |
| CMTM3 | |
| 9430096L06Rik; AI413895; BNAS2; chemokine-like factor super family 3; chemokine-like factor superfamily member 3; CKLF like MARVEL transmembrane domain containing 3; CKLF-like MARVEL transmembrane domain containing 3; CKLF-like MARVEL transmembrane domain-containing protein 3; CKLFSF3; CMTM3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 123920 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96MX0 | |
| CMTM3 | |
| Recombinant protein corresponding to Human CMTM3. Recombinant protein control fragment (Product #RP-90797). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction