missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMTM5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31149
This item is not returnable.
View return policy
Description
CMTM5 Polyclonal specifically detects CMTM5 in Human samples. It is validated for Western Blot.
Specifications
| CMTM5 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| CMTM5 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human CKLFSF5. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.06mg/mL | |
| Western Blot 1:500-1:3000 | |
| chemokine-like factor super family 5, chemokine-like factor superfamily 5, Chemokine-like factor superfamily member 5, CKLF-like MARVEL transmembrane domain containing 5, CKLF-like MARVEL transmembrane domain-containing protein 5, CKLFSF5, FLJ37521 | |
| Rabbit | |
| 25 kDa | |
| 0.1 mL | |
| Cytokine Research | |
| 116173 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction