missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CMTM6 Monoclonal Antibody (RCT11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549503
This item is not returnable.
View return policy
Description
This product was used in an Indirect ELISA.
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. Han W., Genomics 81:609-617(2003). Ota T., Nat. Genet. 36:40-45(2004). The MGC Project Team; Genome Res. 14:2121-2127(2004).
Specifications
| CMTM6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9NX76 | |
| CMTM6 | |
| CMTM6 C-terminal peptide. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| RCT11 | |
| Unconjugated | |
| CMTM6 | |
| 2810051A14Rik; AA536733; chemokine-like factor super family 6; chemokine-like factor superfamily 6; chemokine-like factor superfamily member 6; CKLF like MARVEL transmembrane domain containing 6; CKLF-like MARVEL transmembrane domain containing 6; CKLF-like MARVEL transmembrane domain-containing protein 6; Cklfsf6; Cmtm6; Da2-17; LRRGT00102; PRO2219 | |
| Mouse | |
| Protein A | |
| RUO | |
| 54918 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction