Learn More
IBA Lifesciences CMV pp65 peptide TPRVTGGGAM (HLA-B*0702)
GREENER_CHOICE
Product Code. 15342156 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 15342156
Brand: IBA Lifesciences 67027901
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Single peptide synthesized as presented by the MHC class I allele
  • CMV pp65 peptide TPRVTGGGAM (HLA-B*0702) is a single peptide for stimulation of T cells.
  • The peptide from human cytomegalovirus (CMV) phosphoprotein (pp65) is synthesized as it is presented by the MHC class I HLA-B*0702 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
  • The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
  • Specifications
    Content And Storage Recommended: -25°C to -15°C
    Quantity 1 mg