missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CMYA5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17874810
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17874810 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17874810 Supplier Invitrogen™ Supplier No. PA5140296

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: FISEVSREDY GKKEISGDSE EMNINSVVTS ADGENLEIQS YSLIGEKLVM EEAKTIVPPH VTDSKRVQKP AIAPPSK Highest antigen sequence indentity to the following orthologs: Rat - 48%, Mouse - 40%.

SPRYD2, also known as Myospryn, was originally identified as the muscle-specific partner of dysbindin and as a Mef-2 target gene. It is a large scaffolding protein localized to the Z-disc/costamere region of striated muscle. SPRYD2 includes a noncanonical tripartite motif (TRIM-like) that lacks the RING domain but consists of a B-box coiled coil (BBC), fibronectin 3 (FN3) repeats, and SPRY domains. SPRYD2 interacts with desmin and calcineurin, and it has been suggested to play a role in the biogenesis of lysosome and negatively regulates slow-fiber-type transformation and skeletal muscle regeneration. SPRYD2 is dysregulated in Duchenne muscular dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMYA5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CMYA5
Gene Accession No. Q8N3K9
Gene Alias 2310076E16Rik; 2310076E21Rik; AA420382; AI504003; C5orf10; cardiomyopathy associated 5; cardiomyopathy-associated protein 5; CMYA5; DTNBP2; Dystrobrevin-binding protein 2; genethonin-3; heart and skeletal muscle-specific and sprouty domain-containing; myospryn; SPRY domain-containing protein 2; SPRYD2; sr553; Srfsd; stretch response protein 553; stretch-response protein 553; stretch-responsive fibronectin protein type 3; Tims; TRIM76; TRIM-like protein; tripartite motif-containing 76; tripartite motif-containing protein 76
Gene Symbols CMYA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CMYA5. Recombinant protein control fragment (Product #RP-109651).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202333
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.