missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CN079 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563927

Product Code. 15791692

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SRCQENFFLV LGIDAAQKNL SGGQGHIMED CDLKEPEGLL TVSSFCLQHC KALIQTKLSG PPGSKQGRLM TCSRFLKTPS CGGGQHITIP RKRMFTPRKL Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 66%.

C14orf79 gene ontology annotations related to this gene include intracellular transport.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CN079
Polyclonal
Unconjugated
CLBA1
C14orf79; chromosome 14 open reading frame 79; Clathrin-binding box of aftiphilin-containing protein 1; CLBA1; uncharacterized protein C14orf79; Uncharacterized protein CLBA1
Rabbit
Antigen affinity chromatography
RUO
122616
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96F83
CLBA1
Recombinant protein corresponding to Human CN079. Recombinant protein control fragment (Product #RP-104250).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.