missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNDP1 Monoclonal Antibody (OTI2F8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524927
This item is not returnable.
View return policy
Description
CNDP1 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.
Specifications
| CNDP1 | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96KN2 | |
| CNDP1 | |
| Full length human recombinant protein of CNDP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2F8 | |
| Unconjugated | |
| CNDP1 | |
| AI746433; beta-Ala-His dipeptidase; carnosinase 1; carnosine dipeptidase 1; carnosine dipeptidase 1 (metallopeptidase M20 family); CN1; CNDP dipeptidase 1; CNDP1; CPGL2; glutamate carboxypeptidase-like protein 2; HsT2308; serum carnosinase; UNQ1915/PRO4380 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84735 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction