missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNDP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110757
This item is not returnable.
View return policy
Description
Immunogen sequence: PALLEKVFQY IDLHQDEFVQ TLKEWVAIES DSVQPVPRFR QELFRMMAVA ADTLQRLGAR VASVDMGPQQ LPDGQSLPIP PVILAELGSD PTKGTVCFYG HL Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 80%.
This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.
Specifications
| CNDP1 | |
| Polyclonal | |
| Unconjugated | |
| CNDP1 | |
| AI746433; beta-Ala-His dipeptidase; carnosinase 1; carnosine dipeptidase 1; carnosine dipeptidase 1 (metallopeptidase M20 family); CN1; CNDP dipeptidase 1; CNDP1; CPGL2; glutamate carboxypeptidase-like protein 2; HsT2308; serum carnosinase; UNQ1915/PRO4380 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84735 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96KN2 | |
| CNDP1 | |
| Recombinant Protein corresonding to Human CNDP1. Recombinant protein control fragment (Product #RP-89927). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction