Learn More
Invitrogen™ CNDP1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5121107
Product Code. 17752504
  • € 514.00 / 100µL
Description
Positive test controls include: 293T, A-431, HepG2, Mouse kidney, Mouse brain, Rat spinal cord, Rat brain. The target is usually found in the following locations: Secreted. Immunogen sequence: MDPKLGRMAA SLLAVLLLLL ERGMFSSPSP PPALLEKVFQ YIDLHQDEFV QTLKEWVAIE SDSVQPVPRF RQELFRMMAV AADTLQRLGA RVASVDMGPQ QLPDGQSLPI PPVILAELGS DPTKGTVCFY GHLDVQPADR GDGWLTDPYV LTEVDGKLYG RGATDNKGPV LAWINAVSAF RALEQDLPVN IKFIIEGMEE AGSVALEELV EKEKDRFFSG VDYIVISDNL WISQRKPAIT YGTRGNSYFM VEVKCRDQDF HSGTFGGILH EPMADLVALL.
This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CNDP1
Polyclonal
Unconjugated
CNDP1
AI746433; beta-Ala-His dipeptidase; carnosinase 1; carnosine dipeptidase 1; carnosine dipeptidase 1 (metallopeptidase M20 family); CN1; CNDP dipeptidase 1; CNDP1; CPGL2; glutamate carboxypeptidase-like protein 2; HsT2308; serum carnosinase; UNQ1915/PRO4380
Rabbit
Affinity Chromatography
RUO
307212, 338403, 84735
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.77 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q66HG3, Q8BUG2, Q96KN2
CNDP1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human CNDP1 (NP_116038.4).
100 μL
Primary
Human, Mouse, Rat