missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNDP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17759-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.Specifications
| CNDP1 | |
| Polyclonal | |
| Unconjugated | |
| CNDP1 | |
| Beta Ala His dipeptidase, Carnosine dipeptidase 1, CN1, CNDP dipeptidase 1, CNDP1, CPGL2, HsT2308, Serum carnosinase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 307212, 338403, 84735 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q66HG3, Q8BUG2, Q96KN2 | |
| Cndp1 | |
| CNDP1 Fusion Protein Ag12127 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction