missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNGA4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 462.00
Specifications
| Antigen | CNGA4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CNGA4 Polyclonal antibody specifically detects CNGA4 in Human, Rat samples. It is validated for Western BlotSpecifications
| CNGA4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 1262 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| CNCA2, CNG channel alpha-4, CNG4, CNG-4, CNG5, CNGB2, cyclic nucleotide gated channel alpha 4, cyclic nucleotide gated channel beta 2, cyclic nucleotide-gated cation channel alpha-4, Cyclic nucleotide-gated channel alpha-4, MGC126168, MGC126169, OCNC2, OCNCb, OCNCBETA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 416-575 of human CNGA4 (NP_001032406.1). ISIINIKGNMSGNRRTANIKSLGYSDLFCLSKEDLREVLSEYPQAQTIMEEKGREILLKMNKLDVNAEAAEIALQEATESRLRGLDQQLDDLQTKFARLLAELESSALKIAYRIERLEWQTREWPMPEDLAEADDEGEPEEGTSKDEEGRASQEGPPGPE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title