missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNGB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112936
This item is not returnable.
View return policy
Description
CNGB1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Glutamic acid rich protein (GARP) is a soluble protein localized to the outer segments of the rod photoreceptor. GARPs are characterized by six repeated motifs of: V V E K K N/E E. These repeat motifs help form an alpha-helix structure. The function(s) of GARP have not been fully characterized, but studies have suggested that GARP helps limit cGMP turnover by inhibiting phosphodiesterase activity. To date, there have been two splice variants characterized: GARP1 (full-length GARP) and GARP2 (truncated GARP). GARP also interacts with part of the larger cytoplasmic domain of the beta-subunit of the cyclic GMP-gated channel found in the plasma membrane.
Specifications
| CNGB1 | |
| Polyclonal | |
| Unconjugated | |
| CNGB1 | |
| 240 kDa protein of rod photoreceptor CNG-channel; 240K protein of rod photoreceptor cng-channel; CNCG2; CNCG3L; CNCG4; cng channel; CNG channel 4; CNG channel beta-1; CNG4; CNG-4; CNG4c protein; CNGB1; CNGB1B; cyclic nucleotide gated channel beta 1; cyclic nucleotide gated channel subunit beta 1; cyclic nucleotide-gated cation channel 4; Cyclic nucleotide-gated cation channel beta-1; cyclic nucleotide-gated cation channel gamma; Cyclic nucleotide-gated cation channel modulatory subunit; cyclic nucleotide-gated channel beta subunit 1; cyclic nucleotide-gated channel beta subunit 1c; cyclic nucleotide-gated channel beta subunit 1d; cyclic nucleotide-gated channel beta subunit 1e; cyclic nucleotide-gated channel beta-1; GAR1; GARP; GARP2; glutamic acid-rich protein; glutamic-acid-rich protein; Gm1959; RCNC2; RCNCb; RCNCbeta; RP45; splice variant; tGARP; glutamic acid rich protein; truncated rod photoreceptor cGMP-gated channel beta-subunit | |
| Rabbit | |
| Protein G | |
| RUO | |
| 1258 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q14028 | |
| CNGB1 | |
| Recombinant Human Cyclic nucleotide-gated cation channel beta-1 protein (131-253AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction