missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNKSR3 Antibody (OTI1D7), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72428PCP
This item is not returnable.
View return policy
Description
CNKSR3 Monoclonal antibody specifically detects CNKSR3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| CNKSR3 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1D7 | |
| Western Blot, Immunohistochemistry | |
| CNK homolog protein 3, CNK3, CNKSR family member 3MAGI1, FLJ31349, membrane associated guanylate kinase interacting protein-like 1, membrane-associated guanylate kinase-interacting protein-like 1, WW and PDZ domain containing 1 | |
| Full length human recombinant protein of human CNKSR3 (NP_775786) produced in HEK293T cell. | |
| 0.1 mL | |
| Neuroscience | |
| 154043 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction