missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CNKSR3 Monoclonal Antibody (OTI1D7)
GREENER_CHOICE

Product Code. 15955564
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15955564 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15955564 Supplier Invitrogen™ Supplier No. MA525968

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CNKSR3 Monoclonal Antibody for Western Blot, IHC (P)

CNKSR3 may be involved in transepithelial sodium transport. Regulates aldosterone-induced and ENaC-mediated sodium transport possibly through regulation of the ERK pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNKSR3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CNKSR3
Gene Accession No. Q6P9H4
Gene Alias 6820402C05; BC024086; CNK homolog protein 3; CNK3; CNKSR family member 3; Cnksr3; Connector enhancer of kinase suppressor of ras 3; connector enhancer of KSR 3; KOX14; MAGI1; maguin-like protein; membrane associated guanylate kinase interacting protein-like 1; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase-interacting protein-like 1; parturition-related protein 4; Prp4; ZNF21
Gene Symbols CNKSR3
Host Species Mouse
Immunogen Full length human recombinant protein of CNKSR3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154043
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.