missing translation for 'onlineSavingsMsg'
Learn More

CNKSR3 Monoclonal Antibody (OTI1D7), TrueMAB™, OriGene

Product Code. p-200012497
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17582302 30 μL 30µL
17562302 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17582302 Supplier Origene Technologies Supplier No. TA504737S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CNKSR3 Monoclonal antibody specifically detects CNKSR3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CNKSR3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TBCC
Gene Accession No. Q15814, Q8VCN9
Gene Alias 6820402C05; BC024086; CNK homolog protein 3; CNK3; CNKSR family member 3; Cnksr3; Connector enhancer of kinase suppressor of ras 3; connector enhancer of KSR 3; KOX14; MAGI1; maguin-like protein; membrane associated guanylate kinase interacting protein-like 1; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase-interacting protein-like 1; parturition-related protein 4; Prp4; ZNF21
Host Species Mouse
Immunogen Full length human recombinant protein of human TBCC produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316221, 6903, 72726
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.