missing translation for 'onlineSavingsMsg'
Learn More

CNN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16818643 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16818643 20 μL 20µL
16808643 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16818643 Supplier Proteintech Supplier No. 115091AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a protein with a markedly acidic C terminus; the basic N-terminus is highly homologous to the N-terminus of a related gene, CNN1. Members of the CNN gene family all contain similar tandemly repeated motifs. This encoded protein is associated with the cytoskeleton but is not involved in contraction.

Specifications

Antigen CNN3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CNN3
Gene Accession No. P37397, Q15417, Q9DAW9
Gene Alias Calponin 3, calponin 3, acidic, Calponin, acidic isoform, CNN3
Gene Symbols Cnn3
Host Species Rabbit
Immunogen CNN3 Fusion Protein Ag2068
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1266, 54321, 71994
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.