missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15928
This item is not returnable.
View return policy
Description
CNOT3 Polyclonal specifically detects CNOT3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CNOT3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| CCR4-NOT transcription complex, subunit 3, KIAA0691CCR4-associated factor 3, LENG2yeast) homolog, Leukocyte receptor cluster member 2 | |
| Rabbit | |
| 82 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (98%), Rabbit (100%), Rat (98%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.63mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| O75175 | |
| CNOT3 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human CNOT3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 4849 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction