missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNOT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100231
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CNOT3.
Component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. May be involved in metabolic regulation; may be involved in recruitment of the CCR4-NOT complex to deadenylation target mRNAs involved in energy metabolism. Involved in mitotic progression and regulation of the spindle assembly checkpoint by regulating the stability of MAD1L1 mRNA. Can repress transcription and may link the CCR4-NOT complex to transcriptional regulation; the repressive function may involve histone deacetylases. Involved in the maintenance of embryonic stem (ES) cell identity. [UniProt].
Specifications
| CNOT3 | |
| Polyclonal | |
| Unconjugated | |
| CNOT3 | |
| A930039N10Rik; CCR4-associated factor 3; CCR4-NOT transcription complex subunit 3; CCR4-NOT transcription complex, subunit 3; CNOT3; KIAA0691; LENG2; Leukocyte receptor cluster member 2; Not3; NOT3 (negative regulator of transcription 3, yeast) homolog; NOT3H | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 232791, 308311, 4849 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75175, Q8K0V4 | |
| CNOT3 | |
| A synthesized peptide derived from human CNOT3(Accession O75175), corresponding to amino acid residues S258-V308. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction