missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT4 Antibody (OTI3D11), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71203MFV500
This item is not returnable.
View return policy
Description
CNOT4 Monoclonal antibody specifically detects CNOT4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| CNOT4 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 4850 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI3D11 | |
| Flow Cytometry | |
| CCR4-associated factor 4, CCR4-NOT transcription complex, subunit 4, E3 ubiquitin-protein ligase CNOT4, EC 6.3.2.-, NOT4yeast) homolog | |
| Human recombinant protein fragment corresponding to ammino acids 190-455 of human CNOT4(NP_037448) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction