missing translation for 'onlineSavingsMsg'
Learn More

CNOT4 Monoclonal Antibody (OTI4C7), TrueMAB™, OriGene

Product Code. p-200011595
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17541742 30 μL 30µL
17521742 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17541742 Supplier Origene Technologies Supplier No. TA504475S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CNOT4 Monoclonal antibody specifically detects CNOT4 in Human samples. It is validated for Western Blot

Specifications

Antigen CNOT4
Applications Western Blot
Classification Monoclonal
Clone OTI4C7
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CNOT4
Gene Accession No. O95628
Gene Alias CCR4-associated factor 4; CCR4-NOT transcription complex subunit 4; CCR4-NOT transcription complex subunit 4 L homeolog; CCR4-NOT transcription complex subunit 4 S homeolog; CCR4-NOT transcription complex subunit 4 S homeolog; CCR4-NOT transcription complex, subunit 4; CCR4-NOT transcription complex, subunit 4 isoform a; CLONE243; cnot4; cnot4.S; E3 ubiquitin-protein ligase CNOT4; hypothetical protein; negative regulator of transcription 4; Not4; NOT4 (negative regulator of transcription 4, yeast) homolog; Not4h; Not4hp; Potential transcriptional repressor NOT4Hp; RING-type E3 ubiquitin transferase CNOT4; transcriptional repressor NOT4Hp; XELAEV_18019807mg
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 190-455 of human CNOT4 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4850
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.