missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNOT8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100445
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CNOT8.
CNOT8 (CCR4-NOT transcription complex subunit 8), also known as CALIF or POP2, is a 292 amino acid protein that localizes to both the nucleus and the cytoplasm and functions as part of the CCR-NOT complex. Expressed ubiquitously, CNOT8 plays a role in transcriptional regulation for a diverse set of processes.
Specifications
| CNOT8 | |
| Polyclonal | |
| Unconjugated | |
| CNOT8 | |
| 1500015I04Rik; 1810022F04Rik; AA536816; AU015770; AU043059; CAF1; Caf1b; CAF1-like protein; CAF2; CALIF; CALIFp; CCR4-associated factor 8; CCR4-NOT transcription complex subunit 8; CCR4-NOT transcription complex, subunit 8; CNOT8; h CAF1-like protein; hCAF1; PGK promoter directed over production; POP2; wu:fe49a05; zgc:63844 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 363603, 69125, 9337 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9D8X5, Q9UFF9 | |
| CNOT8 | |
| A synthesized peptide derived from human CNOT8(Accession Q9UFF9), corresponding to amino acid residues P89-L139. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction