missing translation for 'onlineSavingsMsg'
Learn More

CNPase Antibody (CL2871) [Alexa Fluor™ 350], Novus Biologicals™

Product Code. 30523717 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30523717 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30523717 Supplier Novus Biologicals Supplier No. NBP343809AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CNPase Monoclonal antibody specifically detects CNPase in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNPase
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL2871
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias 2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of P09543, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Glia Markers, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1267
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.