Learn More
CNPase Antibody (CL2871) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46635UV
Product Code. 30304719
  • € 737.00 / 0.10mL
Description
CNPase Monoclonal antibody specifically detects CNPase in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CNPase
Monoclonal
DyLight 350
2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37
This antibody was developed using a recombinant protein derived from P09543, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cellular Markers, Glia Markers, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers
1267
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL2871
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat