missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNPase Antibody (CL2872), Novus Biologicals™
Mouse Monoclonal Antibody
€ 369.00 - € 500.00
Specifications
| Antigen | CNPase |
|---|---|
| Clone | CL2872 |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603406
|
Novus Biologicals
NBP2-46636-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676775
|
Novus Biologicals
NBP2-46636 |
0.1 mL |
€ 500.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CNPase Monoclonal antibody specifically detects CNPase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CNPase | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| P09543 | |
| 1267 | |
| IgG1 | |
| Protein A purified |
| CL2872 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Glia Markers, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers | |
| PBS (pH 7.2), 40% Glycerol | |
| 2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence PGVLHCTTKFCDYGKAPGAEEYAQQDVLKKSYSKAFTLTISALFVTPKTTGARVELSEQQLQLWPSDVDKLSPTDNLPRGSRAHITL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title