missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNPase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143601
This item is not returnable.
View return policy
Description
CNPase Polyclonal Antibody for Western Blot, ICC/IF, IHC
CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.
Specifications
| CNPase | |
| Polyclonal | |
| Unconjugated | |
| CNP | |
| 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; Cnp1; Cnp-1; CNPase; CNPF; CNPI; CNPII; cyclic nucleotide phosphodiesterase 1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with 5mM sodium azide | |
| P09543, P13233, P16330 | |
| CNP | |
| Full length human recombinant protein expressed in E. coli. | |
| RUO | |
| 1267, 12799, 25275 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction