missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNPase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA146433
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and rat based on 94% sequence homology. Suggested positive control: antigen standard for CNP (transient overexpression lysate), normal human brain lysate.
CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.
Specifications
| CNPase | |
| Polyclonal | |
| Unconjugated | |
| CNP | |
| 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; Cnp1; Cnp-1; CNPase; CNPF; CNPI; CNPII; cyclic nucleotide phosphodiesterase 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1267, 12799 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P09543, P16330 | |
| CNP | |
| Residues 68-84 of the 46-48 (doublet) kDa human, CNPase protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction