missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cnpy4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555233
This item is not returnable.
View return policy
Description
Immunogen sequence: KEEDDDTERL PSKCEVCKLL STELQAELSR TGRSREVLEL GQVLDTGKRK RHVPYSVSET RLEEALENLC ERI Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Specifications
| Cnpy4 | |
| Polyclonal | |
| Unconjugated | |
| CNPY4 | |
| 2610019P18Rik; canopy 4 homolog; canopy 4 homolog (zebrafish); canopy FGF signaling regulator 4; Cnpy; Cnpy4; Prat4b; PRotein Associated with Tlr4; Protein canopy homolog 4; PSEC0237; RGD1307636; UNQ1909/PRO4354 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 245812 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N129 | |
| CNPY4 | |
| Recombinant protein corresponding to Human Cnpy4. Recombinant protein control fragment (Product #RP-90414). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction