missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTF Antibody (121), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90839IR
This item is not returnable.
View return policy
Description
CNTF Monoclonal specifically detects CNTF in Rat samples. It is validated for ELISA.
Specifications
| CNTF | |
| Monoclonal | |
| DyLight 755 | |
| ciliary neurotrophic factor, HCNTF | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Rat CNTF (P20294; Met1-Met200). | |
| 0.1 mL | |
| Biologically Active Proteins, Neuroscience | |
| 1270 | |
| Rat | |
| Purified |
| ELISA | |
| 121 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human CNTF Mouse LIF Mouse OSM Rat IL6 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction