missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTFR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 463.00
Specifications
| Antigen | CNTFR |
|---|---|
| Dilution | Western Blot 1:400-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667430
|
Novus Biologicals
NBP2-92414-0.02ml |
0.02 mL |
€ 204.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631421
|
Novus Biologicals
NBP2-92414-0.1ml |
0.1 mL |
€ 463.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CNTFR Polyclonal antibody specifically detects CNTFR in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CNTFR | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Growth and Development, Neuronal Cell Markers | |
| PBS with 50% glycerol, pH7.3. | |
| 1271 | |
| IgG | |
| Affinity purified |
| Western Blot 1:400-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ciliary neurotrophic factor receptor, ciliary neurotrophic factor receptor alpha, ciliary neurotrophic factor receptor subunit alpha, CNTF receptor subunit alpha, CNTFR alpha, CNTFR-alpha, MGC1774 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 90-342 of human CNTFR (NP_671693.1). FHRDSWHLRHQVLLHVGLPPREPVLSCRSNTYPKGFYCSWHLPTPTYIPNTFNVTVLHGSKIMVCEKDPALKNRCHIRYMHLFSTIKYKVSISVSNALGHNATAITFDEFTIVKPDPPENVVARPVPSNPRRLEVTWQTPSTWPDPESFPLKFFLRYRPLILDQWQHVELSDGTAHTITDAYAGKEYIIQVAAKDNEIGTWSDWSVAAHATPWTEEPRHLTTEAQAAETTTSTTSSLAPPPTTKICDPGELGS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title