missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTFR Antibody (MM0190-10L29), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12196APC
This item is not returnable.
View return policy
Description
CNTFR Monoclonal antibody specifically detects CNTFR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CNTFR | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0190-10L29 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ciliary neurotrophic factor receptor, ciliary neurotrophic factor receptor alpha, ciliary neurotrophic factor receptor subunit alpha, CNTF receptor subunit alpha, CNTFR alpha, CNTFR-alpha, MGC1774 | |
| Human recombinant CNTFR | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers | |
| 1271 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction