Learn More
CNTFR Antibody (MM0190-10L29), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP2-12196CL1
Product Code. 30155645
  • € 1020.00 / 0.10mL
Description
CNTFR Monoclonal antibody specifically detects CNTFR in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CNTFR
Monoclonal
CoraFluor 1
ciliary neurotrophic factor receptor, ciliary neurotrophic factor receptor alpha, ciliary neurotrophic factor receptor subunit alpha, CNTF receptor subunit alpha, CNTFR alpha, CNTFR-alpha, MGC1774
Human recombinant CNTFR
0.1 mL
Growth and Development, Neuronal Cell Markers
1271
Store at 4°C in the dark. Do not freeze.
IgG2
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
MM0190-10L29
PBS
Mouse
Protein G purified