Learn More
Invitrogen™ CNTFR Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA547437
Description
In Western blots, greater than 50% cross-reactivity with recombinant rat CNTF sR alpha is observed and less than 5% cross-reactivity with recombinant human (rh) IL-2 sR gamma and rhIL-6 sR is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
This gene encodes a hematopoeitin/interferon-class receptor belonging to the cytokine superfamily of receptors. The encoded gene product represents the CNTF-specific alpha subunit of a heterotrimer forming the CNTF receptor complex, which also includes LIFR and gp130. The receptor is attached to the membrane by a glycosyl-phosphatidylinositol linkage and contains an immunoglobulin-like C2-type domain and a fibronectin type-III domain. Signal transduction requires that CNTF bind first to this alpha component, which permits the recruitment of gp130 and LIFR beta to form the tripartite receptor complex. Signal transduction stimulates gene expression, cell survival or differentiation in a variety of neuronal cell types. Alternative splicing has been observed at this locus and two variants, both encoding the same protein, have been identified.
Specifications
| CNTFR | |
| Polyclonal | |
| Unconjugated | |
| Cntfr | |
| ciliary neurotrophic factor receptor; ciliary neurotrophic factor receptor alpha; ciliary neurotrophic factor receptor subunit alpha; CNTF receptor subunit alpha; Cntfr; CNTFR alpha; CNTFRALPHA; CNTFR-alpha; MGC1774 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 1271 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P26992 | |
| Cntfr | |
| Sf 21-derived recombinant human CNTF R alpha Gln23-Pro346 (predicted). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.