missing translation for 'onlineSavingsMsg'
Learn More

COA6 Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-06320CL1

Product Code. 30155578

  • € 822.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

COA6 Polyclonal antibody specifically detects COA6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

COA6
Polyclonal
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
388753
Store at 4°C in the dark. Do not freeze.
IgG
Immunocytochemistry/Immunofluorescence
CoraFluor 1
chromosome 1 open reading frame 31, cytochrome c oxidase assembly factor 6 homolog (S. cerevisiae), hypothetical protein LOC388753
Produced in rabbits immunized with E. coli-derived Human COA6 fragment.
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.