missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COA6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555908
This item is not returnable.
View return policy
Description
Immunogen sequence: PSMKERQVCW GARDEYWKCL DENLEDASQC KKLRSSFESS CPQQWIKYFD KRRDYLKFKE KFEAGQFEPS ETTA Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 76%.
C1orf31, also known as COA6, is a conserved mitochondrial protein required for respiratory complex IV biogenesis. Recently study reported that C1orf31 is required for maintenance of cytochrome c oxidase (CcO) subunit levels, and has role in the Cu delivery pathway to CcO. Defects in C1orf31 is associated with mitochondrial respiratory chain disease (MRCD). (24549041).
Specifications
| COA6 | |
| Polyclonal | |
| Unconjugated | |
| COA6 | |
| 1810063B05Rik; AI447995; C1orf31; ca031; CEMCOX4; Coa6; cytochrome c oxidase assembly factor 6; cytochrome c oxidase assembly factor 6 homolog; LOC102556031; uncharacterized LOC102556031 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 388753 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5JTJ3 | |
| COA6 | |
| Recombinant protein corresponding to Human COA6. Recombinant protein control fragment (Product #RP-94445). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction