missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coactosin-like Protein 1/CotL1 Antibody (1D6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-31203-0.1ML
This item is not returnable.
View return policy
Description
Coactosin-like Protein 1/CotL1 Monoclonal specifically detects Coactosin-like Protein 1/CotL1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Coactosin-like Protein 1/CotL1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA 1:1000 | |
| CLPFLJ43657, coactosin-like 1 (Dictyostelium), coactosin-like protein, MGC19733 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 23406 | |
| Protein A purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA | |
| 1D6 | |
| Unconjugated | |
| PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide | |
| COTL1 | |
| Recombinant human COTL1 (1-142aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Anti-human COTL1 mAb, clone 1D6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human COTL1 protein. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido