missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coactosin-like Protein 1/CotL1 Antibody (5C8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37519
This item is not returnable.
View return policy
Description
Coactosin-like Protein 1/CotL1 Monoclonal specifically detects Coactosin-like Protein 1/CotL1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Coactosin-like Protein 1/CotL1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| CLPFLJ43657, coactosin-like 1 (Dictyostelium), coactosin-like protein, MGC19733 | |
| Mouse | |
| 16 kDa | |
| 0.1 mL | |
| Cytokine Research, Signal Transduction | |
| 23406 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 5C8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| COTL1 | |
| Purified recombinant fragment of human COTL1 expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction