missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor VII Antibody (F7/3511), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08251IR
This item is not returnable.
View return policy
Description
Coagulation Factor VII Monoclonal antibody specifically detects Coagulation Factor VII in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Coagulation Factor VII | |
| Monoclonal | |
| DyLight 755 | |
| coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA | |
| Recombinant fragment (around aa 366-466) of human Coagulation Factor VII protein (Uniprot: P08709) | |
| 100 μg | |
| Cardiovascular Biology | |
| 2155 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| ELISA, Immunohistochemistry (Paraffin), Protein Array | |
| F7/3511 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction