missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor XI Antibody (006), Alexa Fluor™ 750 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89239AF750
This item is not returnable.
View return policy
Description
Coagulation Factor XI Monoclonal antibody specifically detects Coagulation Factor XI in Human samples. It is validated for ELISA
Specifications
| Coagulation Factor XI | |
| Monoclonal | |
| Alexa Fluor 750 | |
| coagulation factor XI, EC 3.4.21, EC 3.4.21.27, FXIPlasma thromboplastin antecedent, MGC141891, PTA | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Coagulation Factor XI (NP_000119.1; Met1-Val625). | |
| 0.1 mL | |
| Prostate Cancer | |
| 2160 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 006 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction