missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA9 Monoclonal Antibody (OTI1C6), TrueMAB™, OriGene
Description
MAGEA9 Monoclonal antibody specifically detects MAGEA9 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MAGEA9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1C6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | CD99 |
| Gene Accession No. | P14209 |
| Gene Alias | Cancer/testis antigen 1.9; cancer/testis antigen family 1, member 9; CT1.9; MAGE family member A9; MAGE9; MAGE-9 antigen; MAGEA9; MAGEA9A; MAGEA9B; melanoma antigen family A, 9; melanoma antigen family A9; melanoma-associated antigen 9; MGC114427; similar to melanoma antigen family A, 5; uncharacterized protein LOC317297 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?